This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD248 SIRNA Assays ›
Gene Symbol:
CD248
Gene Name:
CD248 molecule
Gene Aliases:
CD164L1, TEM1
Chromosome Location:
Chr.11: 66314487 - 66317044 on Build GRCh38
UniGene ID:
Hs.195727
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 28396

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD248
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020404.2 NP_065137.1 1 2486
GenBank mRNA AF279142.1 - 1 2474
AJ295846.1 - 1 2486
AK027290.1 - 1 1608
BC051340.1 - 1 2473 6339237
BC098158.1 - 3 180
BC104484.1 - 1 2486 40007855
BC105633.1 - 1 2486 40007854

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
positive regulation of cell proliferation
cell migration
lymph node development
anatomical structure regression
positive regulation of endothelial cell apoptotic process
molecular_function
calcium ion binding
carbohydrate binding
extracellular matrix binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0