This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DYRK2 SIRNA Assays ›
Gene Symbol:
Dyrk2
Gene Name:
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
Gene Aliases:
1810038L18Rik
Chromosome Location:
Chr.10: 118855602 - 118889246 on Build GRCm38
UniGene ID:
Mm.45565
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 283713

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dyrk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014390.2 NP_001014412.1 3 961
GenBank mRNA AK149371.1 - 3 1197
AK155496.1 - 3 1210
BC085145.1 - 3 961 6808145

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
smoothened signaling pathway
phosphorylation
peptidyl-tyrosine phosphorylation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of glycogen biosynthetic process
negative regulation of NFAT protein import into nucleus
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
manganese ion binding
ubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0