siRNA Details
Gene Symbol: Wdr91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013366.1 | NP_001013384.1 | 2 | 241 | ||
GenBank mRNA | AK089347.1 | - | 1 | 1401 | ||
BC055046.1 | - | 2 | 241 | 6488548 |

Gene Symbol: Wdr91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013366.1 | NP_001013384.1 | 2 | 241 | ||
GenBank mRNA | AK089347.1 | - | 1 | 1401 | ||
BC055046.1 | - | 2 | 241 | 6488548 |