siRNA Details
Gene Symbol: Olfr1355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207571.2 | NP_997454.1 | 2 | 637 | ||
GenBank mRNA | AF042360.1 | - | 1 | 76 |

Gene Symbol: Olfr1355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207571.2 | NP_997454.1 | 2 | 637 | ||
GenBank mRNA | AF042360.1 | - | 1 | 76 |