This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MZF1 SIRNA Assays ›
Gene Symbol:
Mzf1
Gene Name:
myeloid zinc finger 1
Gene Aliases:
Mzf-2, Mzf2, Zfp121, Zfp98, Znf42
Chromosome Location:
Chr.7: 13042303 - 13054764 on Build GRCm38
UniGene ID:
Mm.10103
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 282818

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mzf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290453.1 NP_001277382.1 3 812
NM_145819.2 NP_665818.2 3 812
GenBank mRNA AB007407.1 - 2 1031
AB088478.1 - 3 713
AK030410.1 - 3 813
AK145014.1 - 3 693
AK158837.1 - 1 2064
BC060649.1 - 3 772
BC129905.1 - 3 562 40111655

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein homodimerization activity
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header