This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GRAP2 SIRNA Assays ›
Gene Symbol:
Grap2
Gene Name:
GRB2-related adaptor protein 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 121791330 - 121864835 on Build Rnor_6.0
UniGene ID:
Rn.206804
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 282251

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.14
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Grap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034944.1 NP_001030116.1 4 425
GenBank mRNA BC105895.1 - 4 425 7380480

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0