This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CSMD1 SIRNA Assays ›
Gene Symbol:
Csmd1
Gene Name:
CUB and Sushi multiple domains 1
Gene Aliases:
Mdcr1
Chromosome Location:
Chr.16: 77152823 - 78850222 on Build Rnor_6.0
UniGene ID:
Rn.137916
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 282216

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Csmd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037327.1 NP_001032404.1 20 3280
GenBank mRNA DQ124115.1 - 20 3280

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_053171, AK081081, AY017475.

Gene Ontology Categories:

Function(s)

startle response
glucose homeostasis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header