This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARSA SIRNA Assays ›
Gene Symbol:
Arsa
Gene Name:
arylsulfatase A
Gene Aliases:
-
Chromosome Location:
Chr.7: 130448291 - 130450817 on Build Rnor_6.0
UniGene ID:
Rn.23323
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 281977

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arsa
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034933.2 NP_001030105.2 6 1063
GenBank mRNA BC105852.1 - 6 1444 7452956
FQ212178.1 - 2025

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009713, AK004540, BC011284, AK165921, AK132501, and others.

Gene Ontology Categories:

Function(s) Process(es)

autophagy
binding of sperm to zona pellucida
central nervous system development
response to nutrient
response to pH
response to estrogen
response to ethanol
response to methylmercury
arylsulfatase activity
cerebroside-sulfatase activity
calcium ion binding
sulfuric ester hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header