This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF126 SIRNA Assays ›
Gene Symbol:
Rnf126
Gene Name:
ring finger protein 126
Gene Aliases:
-
Chromosome Location:
Chr.7: 12820812 - 12828325 on Build Rnor_6.0
UniGene ID:
Rn.143411
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 281966

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf126
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033702.1 NP_001028874.1 4 343
XM_006240899.1 XP_006240961.1 4 293
GenBank mRNA BC099810.1 - 4 343 5621533

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein monoubiquitination
regulation of protein localization
negative regulation of epidermal growth factor receptor signaling pathway
regulation of cell proliferation
retrograde transport, endosome to Golgi
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
protein K63-linked ubiquitination
protein K48-linked ubiquitination
cytoplasm-associated proteasomal ubiquitin-dependent protein catabolic process
epidermal growth factor receptor binding
zinc ion binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0