This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COX6A1 SIRNA Assays ›
Gene Symbol:
Cox6a1
Gene Name:
cytochrome c oxidase, subunit VIa, polypeptide 1
Gene Aliases:
COX6AL
Chromosome Location:
Chr.12: 47024442 - 47027495 on Build Rnor_6.0
UniGene ID:
Rn.880
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 281304

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cox6a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012814.1 NP_036946.1 3 439
GenBank mRNA AA685532.1 - 1 146
CB716915.1 - 3 439
FQ211611.1 - 3 426
FQ213920.1 - 3 426
FQ221129.1 - 3 431
FQ221354.1 - 3 426
FQ221968.1 - 3 428
FQ224366.1 - 3 426
FQ225833.1 - 3 426
X12553.1 - 3 340

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hydrogen ion transmembrane transport
cytochrome-c oxidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0