This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXW5 SIRNA Assays ›
Gene Symbol:
Fbxw5
Gene Name:
F-box and WD repeat domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 2726735 - 2731213 on Build Rnor_6.0
UniGene ID:
Rn.136952
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 281110

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxw5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025730.2 NP_001020901.1 7, 8 1389
XM_006233623.2 XP_006233685.1 6, 7 1748
XM_006233624.2 XP_006233686.1 8, 9 1451
XM_006233625.2 XP_006233687.1 8, 9 1693
XM_006233626.2 XP_006233688.1 7, 8 1345
XM_006233627.2 XP_006233689.1 7, 8 1589
XM_006233628.2 XP_006233690.1 6, 7 1105
GenBank mRNA BC099179.1 - 7, 8 1392 7383052

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013908, AK154911, AF176520, BC010776, AK188841, and others.

Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
centrosome duplication
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0