This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DHTKD1 SIRNA Assays ›
Gene Symbol:
Dhtkd1 LOC103694860
Gene Name:
dehydrogenase E1 and transketolase domain containing 1
probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial
Gene Aliases:
-
-
Chromosome Location:
Chr.17: 76306585 - 76358058 on Build Rnor_6.0
Chromosome Location:
Chr.Un|NW_007906797: 2461 - 4602 on Build Rnor_6.0
UniGene ID:
Rn.13539
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 281080

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dhtkd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025720.1 NP_001020891.1 11 2091
GenBank mRNA BC099075.1 - 11 2091 7441857
Gene Symbol: LOC103694860
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773802.1 XP_008772024.1 1 197

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081131, BC117994, AK049272, AK173223, AK050057.

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
generation of precursor metabolites and energy
glycolytic process
tricarboxylic acid cycle
oxoglutarate dehydrogenase (succinyl-transferring) activity
thiamine pyrophosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0