This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM166A SIRNA Assays ›
Gene Symbol:
Fam166a
Gene Name:
family with sequence similarity 166, member A
Gene Aliases:
-
Chromosome Location:
Chr.3: 2434367 - 2441706 on Build Rnor_6.0
UniGene ID:
Rn.215050
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 280991

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam166a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025694.2 NP_001020865.1 7 809
XM_008761589.1 XP_008759811.1 8 792
XM_008761590.1 XP_008759812.1 8 784
XM_008761591.1 XP_008759813.1 7 732
XM_008761592.1 XP_008759814.1 7 608
XM_008761593.1 XP_008759815.1 6 647
XM_008761594.1 XP_008759816.1 5 465
XM_008761595.1 XP_008759817.1 4 411
XM_008761596.1 XP_008759818.1 8 862
GenBank mRNA BC097396.1 - 7 810 7453380

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0