This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACNA1H SIRNA Assays ›
Gene Symbol:
Cacna1h
Gene Name:
calcium channel, voltage-dependent, T type, alpha 1H subunit
Gene Aliases:
-
Chromosome Location:
Chr.10: 14730932 - 14788617 on Build Rnor_6.0
UniGene ID:
Rn.49178
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 280720

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cacna1h
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153814.2 NP_722521.1 16 3703
XM_008767532.1 XP_008765754.1 15 8082
XM_008767533.1 XP_008765755.1 15 8082
XM_008767534.1 XP_008765756.1 15 8082
XM_008767535.1 XP_008765757.1 15 8082
XM_008767536.1 XP_008765758.1 14 3301
GenBank mRNA AF290213.2 - 16 3706

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
sensory perception of pain
aldosterone biosynthetic process
cellular response to hormone stimulus
cortisol biosynthetic process
regulation of ion transmembrane transport
cellular response to potassium ion
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
positive regulation of acrosome reaction
low voltage-gated calcium channel activity
metal ion binding
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0