This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ENY2 SIRNA Assays ›
Gene Symbol:
ENY2
Gene Name:
ENY2, transcription and export complex 2 subunit
Gene Aliases:
DC6, Sus1, e(y)2
Chromosome Location:
Chr.8: 109334323 - 109345960 on Build GRCh38
UniGene ID:
Hs.492555
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 28031

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ENY2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193557.1 NP_001180486.1 3 211
NM_020189.5 NP_064574.1 3 283
NR_036471.1 3 283
NR_036472.1 3 283
GenBank mRNA AF173296.1 - 3 353
AF201940.1 - 3 298
AK000487.1 - 3 187
AK095651.1 - 3 282
AK290752.1 - 3 227
AK316560.1 - 3 278
BC007870.2 - 3 199 4303290
BX648621.1 - 1 1032
CR457183.1 - 2 137
DA548665.1 - 3 157
DA762166.1 - 3 265
DB168034.1 - 3 283

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001130580, XM_006241615, FQ223243.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
protein transport
histone deubiquitination
poly(A)+ mRNA export from nucleus
positive regulation of transcription, DNA-templated
negative regulation of insulin secretion involved in cellular response to glucose stimulus
chromatin binding
transcription coactivator activity
ligand-dependent nuclear receptor transcription coactivator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0