This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CIT SIRNA Assays ›
Gene Symbol:
Cit
Gene Name:
citron rho-interacting serine/threonine kinase
Gene Aliases:
-
Chromosome Location:
Chr.12: 46334669 - 46494155 on Build Rnor_6.0
UniGene ID:
Rn.89069
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 280278

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cit
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029911.1 NP_001025082.1 29 3862
XM_006249469.2 XP_006249531.1 30 3840
XM_006249470.2 XP_006249532.1 30 3840
XM_006249471.2 XP_006249533.1 30 3840
XM_006249472.2 XP_006249534.1 30 3840
XM_006249477.2 XP_006249539.1 20 3028
XM_008769269.1 XP_008767491.1 30 3840
XM_008769270.1 XP_008767492.1 30 3796
XM_008769271.1 XP_008767493.1 29 3866
GenBank mRNA AF039218.1 - 20 2966

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic sister chromatid segregation
G2/M transition of mitotic cell cycle
protein phosphorylation
Golgi organization
mitotic nuclear division
metaphase/anaphase transition of mitotic cell cycle
spermatogenesis
regulation of actin polymerization or depolymerization
phosphorylation
dendrite development
positive regulation of cytokinesis
intracellular signal transduction
generation of neurons
negative regulation of dendrite morphogenesis
regulation of catalytic activity
regulation of cell cycle
protein serine/threonine kinase activity
ATP binding
kinase activity
Rho GTPase binding
PDZ domain binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0