This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM161A SIRNA Assays ›
Gene Symbol:
FAM161A
Gene Name:
family with sequence similarity 161 member A
Gene Aliases:
RP28
Chromosome Location:
Chr.2: 61800240 - 61856074 on Build GRCh38
UniGene ID:
Hs.440466
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 278946

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM161A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201543.1 NP_001188472.1 3 1130
NM_032180.2 NP_115556.2 3 1130
NR_037710.1 3 1074
XM_017005072.1 XP_016860561.1 3 1083
XM_017005073.1 XP_016860562.1 3 1057
XM_017005074.1 XP_016860563.1 3 1030
GenBank mRNA AK023367.1 - 4 1289
AK296255.1 - 2 664
AK310908.1 - 3 1015
BC036448.1 - 3 1028
BC107162.1 - 3 902
BC107163.1 - 3 902
BX648834.1 - 3 1040
BX649029.1 - 3 1041

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

visual perception
cilium assembly
response to stimulus
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0