This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF391 SIRNA Assays ›
Gene Symbol:
ZNF391
Gene Name:
zinc finger protein 391
Gene Aliases:
dJ153G14.3
Chromosome Location:
Chr.6: 27374635 - 27403908 on Build GRCh38
UniGene ID:
Hs.651176
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 274356

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF391
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076781.2 NP_001070249.1 3 490
NM_001322288.1 NP_001309217.1 4 714
NM_001322289.1 NP_001309218.1 4 569
NM_001322293.1 NP_001309222.1 3 458
XM_005249082.3 XP_005249139.1 3 275
XM_006715086.2 XP_006715149.1 3 216
XM_011514569.1 XP_011512871.1 3 198
GenBank mRNA AK294965.1 - 3 449
DA758693.1 - 4 295
DB221901.1 - 3 446

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0