siRNA Details
Gene Symbol: LOC344887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033752.2 | 3 | 580 | |||
GenBank mRNA | AK309325.1 | - | 3 | 580 |

Gene Symbol: LOC344887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033752.2 | 3 | 580 | |||
GenBank mRNA | AK309325.1 | - | 3 | 580 |