siRNA Details
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 9 | 4277 | ||
GenBank mRNA | AJ278245.1 | - | 9 | 3885 | ||
BC070049.1 | - | 9 | 3973 | 30343188 |

Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 9 | 4277 | ||
GenBank mRNA | AJ278245.1 | - | 9 | 3885 | ||
BC070049.1 | - | 9 | 3973 | 30343188 |