This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FRMD4B SIRNA Assays ›
Gene Symbol:
FRMD4B
Gene Name:
FERM domain containing 4B
Gene Aliases:
6030440G05Rik, GRSP1
Chromosome Location:
Chr.3: 69168782 - 69541340 on Build GRCh38
UniGene ID:
Hs.371681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 273356

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FRMD4B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015123.2 NP_055938.1 17 1892
XM_005264722.1 XP_005264779.1 17 1569
XM_005264723.1 XP_005264780.1 7 1051
XM_017005989.1 XP_016861478.1 17 1676
XM_017005990.1 XP_016861479.1 17 1569
XM_017005991.1 XP_016861480.1 18 1729
XM_017005992.1 XP_016861481.1 17 1509
XM_017005993.1 XP_016861482.1 17 1500
XM_017005994.1 XP_016861483.1 17 3342
XM_017005995.1 XP_016861484.1 11 1086
XM_017005996.1 XP_016861485.1 7 788
XM_017005997.1 XP_016861486.1 7 1051
GenBank mRNA AB023230.1 - 17 1661
AK295045.1 - 12 1134
AL832231.1 - 7 889
BC110637.1 - 2 113

Back To Top

More Information


Gene Ontology Categories:

Function(s)

establishment of epithelial cell polarity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0