This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF823 SIRNA Assays ›
Gene Symbol:
ZNF823
Gene Name:
zinc finger protein 823
Gene Aliases:
HSZFP36
Chromosome Location:
Chr.19: 11721265 - 11739009 on Build GRCh38
UniGene ID:
Hs.142167
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 271953

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF823
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080493.3 NP_001073962.1 4 1048
NM_001297610.1 NP_001284539.1 3 987
NM_017507.1 NP_059977.1 3 921
XM_017026941.1 XP_016882430.1 4 1210
XM_017026942.1 XP_016882431.1 2 848
GenBank mRNA AK300579.1 - 4 1015
AK303213.1 - 3 952
BC052597.1 - 4 960 4304302
BC063560.1 - 1 221
X51760.1 - 4 962

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0