This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TET3 SIRNA Assays ›
Gene Symbol:
TET3
Gene Name:
tet methylcytosine dioxygenase 3
Gene Aliases:
hCG_40738
Chromosome Location:
Chr.2: 73982036 - 74108176 on Build GRCh38
UniGene ID:
Hs.516107
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 271560

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TET3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005264187.3 XP_005264244.1 2 524
XM_011532682.2 XP_011530984.1 2 524
XM_011532683.2 XP_011530985.1 2 833
XM_011532684.2 XP_011530986.1 2 2265
XM_011532685.2 XP_011530987.1 2 524
XM_011532686.2 XP_011530988.1 2 524
XM_011532688.2 XP_011530990.1 2 524
XM_017003566.1 XP_016859055.1 2 2364

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein O-linked glycosylation
DNA demethylation of male pronucleus
positive regulation of transcription from RNA polymerase II promoter
oxidation-reduction process
DNA demethylation
histone H3-K4 trimethylation
DNA binding
protein binding
metal ion binding
methylcytosine dioxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0