This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC653080 SIRNA Assays ›
Gene Symbol:
LOC653080 GUSBP2
Gene Name:
glucuronidase beta pseudogene
glucuronidase, beta pseudogene 2
Gene Aliases:
-
GUSBL1, GUSBP4, SMA3-L, SMAC3L, SMAC3L2, b55C20.1, bA239L20.1, bA239L20.5, bGLU-Lp
Chromosome Location:
Chr.5: 70785437 - 70795916 on Build GRCh38
Chromosome Location:
Chr.6: 26871487 - 26956554 on Build GRCh38
UniGene ID:
Hs.582500
Hs.675815
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 270696

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC653080
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA Z70705.1 - 1 74
Gene Symbol: GUSBP2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC067351.1 - 5 682

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0