This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLSCR2 SIRNA Assays ›
Gene Symbol:
PLSCR2
Gene Name:
phospholipid scramblase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 146391363 - 146497163 on Build GRCh38
UniGene ID:
Hs.744414
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 270570

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PLSCR2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011513013.2 XP_011511315.2 8 4693
XM_017006899.1 XP_016862388.1 8 4693
XM_017006901.1 XP_016862390.1 9 4566
XM_017006905.1 XP_016862394.1 10 4149
XM_017006906.1 XP_016862395.1 10 3930
XM_017006907.1 XP_016862396.1 10 3695
XM_017006908.1 XP_016862397.1 10 3820
XM_017006909.1 XP_016862398.1 9 3607
XM_017006912.1 XP_016862401.1 8 3348
XM_017006914.1 XP_016862403.1 8 3555
XM_017006915.1 XP_016862404.1 8 3537

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phospholipid scrambling
calcium ion binding
phospholipid scramblase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm