This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100132147 SIRNA Assays ›
Gene Symbol:
LOC100132147 LOC103091866 LOC440570
Gene Name:
uncharacterized LOC100132147
uncharacterized LOC103091866
uncharacterized LOC440570
Gene Aliases:
-
-
-
Chromosome Location:
Chr.1: 16533891 - 16535652 on Build GRCh38
Chromosome Location:
Chr.1: 145168405 - 145216071 on Build GRCh38
Chromosome Location:
Chr.1|NT_187368: 30201 - 33343 on Build GRCh38
UniGene ID:
Hs.645945
Hs.662922
Hs.308060
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 270210

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100132147
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA HY209018.1 - 1 281
Gene Symbol: LOC103091866
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_111954.1 3 725
NR_111956.1 2 481
GenBank mRNA CA777704.1 - 3 440
Gene Symbol: LOC440570
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AA626381.1 - 1 331
DB338211.1 - 1 318

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0