This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NEMP2 SIRNA Assays ›
Gene Symbol:
NEMP2
Gene Name:
nuclear envelope integral membrane protein 2
Gene Aliases:
TMEM194B
Chromosome Location:
Chr.2: 190454088 - 190607126 on Build GRCh38
UniGene ID:
Hs.659824
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 268854

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NEMP2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142645.1 NP_001136117.1 7 850
NR_136298.1 7 804
XM_005246212.3 XP_005246269.1 7 1004
XM_005246213.3 XP_005246270.1 7 689
XM_011510458.2 XP_011508760.1 7 830
XM_011510459.2 XP_011508761.1 7 1035
XM_017003098.1 XP_016858587.1 7 837
XM_017003099.1 XP_016858588.1 7 837
XM_017003100.1 XP_016858589.1 7 837
GenBank mRNA AK296987.1 - 7 798
AK302425.1 - 6 860

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0