This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC61 SIRNA Assays ›
Gene Symbol:
CCDC61
Gene Name:
coiled-coil domain containing 61
Gene Aliases:
-
Chromosome Location:
Chr.19: 45995457 - 46018616 on Build GRCh38
UniGene ID:
Hs.515479
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 266720

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC61
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267723.1 NP_001254652.1 3 238
XM_005259191.3 XP_005259248.1 3 2343
XM_005259192.4 XP_005259249.1 3 984
XM_011527257.2 XP_011525559.1 3 267
GenBank mRNA DR007040.1 - 3 258
GQ375050.1 - 3 211

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0