siRNA Details
Gene Symbol: LOC102724279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302493.1 | NP_001289422.1 | 2 | 118 | ||
GenBank mRNA | BX095604.1 | - | 2 | 118 | ||
GD139371.1 | - | 2 | 93 |

Gene Symbol: LOC102724279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302493.1 | NP_001289422.1 | 2 | 118 | ||
GenBank mRNA | BX095604.1 | - | 2 | 118 | ||
GD139371.1 | - | 2 | 93 |