This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM150B SIRNA Assays ›
Gene Symbol:
TMEM150B
Gene Name:
transmembrane protein 150B
Gene Aliases:
TMEM224
Chromosome Location:
Chr.19: 55308871 - 55334043 on Build GRCh38
UniGene ID:
Hs.382625
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 265182

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM150B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085488.2 NP_001078957.1 6 483
NM_001282011.1 NP_001268940.1 6 532
NR_104066.1 5 464
NR_104071.1 5 415
XM_005258812.3 XP_005258869.1 8 812
XM_011526850.2 XP_011525152.1 7 691
XM_011526851.2 XP_011525153.1 7 689
XM_011526852.2 XP_011525154.1 7 689
XM_011526853.2 XP_011525155.1 6 603
XM_011526854.2 XP_011525156.1 5 768
XM_011526855.2 XP_011525157.1 7 641
XM_011526856.2 XP_011525158.1 5 720
XM_011526857.2 XP_011525159.1 4 383
XM_017026667.1 XP_016882156.1 7 703
XM_017026668.1 XP_016882157.1 4 406
GenBank mRNA AU311823.1 - 6 483
BC171893.1 - 5 380
BC171901.1 - 5 429
BC171904.1 - 6 493 9054558

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0