This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HAUS5 SIRNA Assays ›
Gene Symbol:
HAUS5
Gene Name:
HAUS augmin like complex subunit 5
Gene Aliases:
KIAA0841, dgt5
Chromosome Location:
Chr.19: 35612744 - 35625349 on Build GRCh38
UniGene ID:
Hs.7426
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 264773

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HAUS5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015302.1 NP_056117.1 15 1343
XM_011526684.2 XP_011524986.1 14 1230
GenBank mRNA AB020648.1 - 15 1316
BC013947.2 - 15 1315
BC064390.1 - 14 1279
BC157882.1 - 15 1334 8862704
BC171905.1 - 15 1335

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
mitotic nuclear division
spindle assembly
centrosome organization
cell division
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0