This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NUP133 SIRNA Assays ›
Gene Symbol:
NUP133
Gene Name:
nucleoporin 133
Gene Aliases:
hNUP133
Chromosome Location:
Chr.1: 229440995 - 229508343 on Build GRCh38
UniGene ID:
Hs.12457
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26459

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NUP133
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018230.2 NP_060700.2 4, 5 613
GenBank mRNA AK001676.1 - 4, 5 613
AK092339.1 - 4, 5 570
AK304061.1 - 5, 6 580
AK304861.1 - 5, 6 514
AK314431.1 - 4, 5 570
BC020107.1 - 4, 5 597 3919973

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery
chromatin organization
regulation of transcription, DNA-templated
mRNA export from nucleus
tRNA export from nucleus
protein import into nucleus
nuclear pore organization
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
neural tube development
neurogenesis
gene silencing by RNA
nuclear pore distribution
paraxial mesoderm development
somite development
intracellular transport of virus
regulation of cellular response to heat
nucleocytoplasmic transporter activity
protein binding
structural constituent of nuclear pore

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0