This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM166B SIRNA Assays ›
Gene Symbol:
FAM166B
Gene Name:
family with sequence similarity 166 member B
Gene Aliases:
-
Chromosome Location:
Chr.9: 35561830 - 35563899 on Build GRCh38
UniGene ID:
Hs.98959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 264059

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM166B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099951.3 NP_001093421.1 3 481
NM_001164310.2 NP_001157782.1 3 481
NM_001287238.1 NP_001274167.1 3 481
NM_001287239.1 NP_001274168.1 3 481
XM_011518028.2 XP_011516330.1 3 620
GenBank mRNA BC129999.1 - 3 481 40127800
BC146928.1 - 3 481 9056856
BC146933.1 - 3 481 9056861
BC171840.1 - 3 481 9054494
BC171851.1 - 3 481 9054505

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0