This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR1689 SIRNA Assays ›
Gene Symbol:
Olr1689 LOC100910420
Gene Name:
olfactory receptor 1689
olfactory receptor 14J1-like
Gene Aliases:
-
-
Chromosome Location:
Chr.20: 933689 - 934660 on Build Rnor_6.0
Chromosome Location:
Chr.20: 752804 - 753873 on Build Rnor_6.0
UniGene ID:
Rn.142688
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 262639

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1689
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000889.1 NP_001000889.1 1 928
Gene Symbol: LOC100910420
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751906.2 XP_003751954.1 945

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0