siRNA Details
Gene Symbol: Olr262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000221.1 | NP_001000221.1 | 1 | 922 |

Gene Symbol: Olr262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000221.1 | NP_001000221.1 | 1 | 922 |