This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCND2 SIRNA Assays ›
Gene Symbol:
Ccnd2
Gene Name:
cyclin D2
Gene Aliases:
-
Chromosome Location:
Chr.4: 159674885 - 159697207 on Build Rnor_6.0
UniGene ID:
Rn.96083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 261243

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccnd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022267.1 NP_071603.1 5 1167
GenBank mRNA L09752.1 - 5 1167

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009829, BC049086, AK147345.

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
ovarian follicle development
liver development
positive regulation of protein phosphorylation
spermatogenesis
positive regulation of cell proliferation
response to organic cyclic compound
response to estradiol
cellular response to insulin stimulus
response to testosterone
response to peptide hormone
response to ethanol
regulation of neuron differentiation
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of epithelial cell proliferation
cell division
response to cAMP
regulation of cell cycle
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0