This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MZF1 SIRNA Assays ›
Gene Symbol:
Mzf1
Gene Name:
myeloid zinc finger 1
Gene Aliases:
Mzf-2, Mzf2, Zfp121, Zfp98, Znf42
Chromosome Location:
Chr.7: 13042303 - 13054764 on Build GRCm38
UniGene ID:
Mm.10103
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 261196

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mzf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290452.1 NP_001277381.1 2 710
NM_001290453.1 NP_001277382.1 2 710
NM_145819.2 NP_665818.2 2 710
GenBank mRNA AB007407.1 - 1 929
AB088478.1 - 2 611
AK030410.1 - 2 711
AK158837.1 - 1 1586
BC060649.1 - 2 670
BC129904.1 - 2 460 40111654
BC129905.1 - 2 460 40111655

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108470, XM_008758767, XM_008758768, XM_006228116.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein homodimerization activity
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0