This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RHOX9 SIRNA Assays ›
Gene Symbol:
Rhox9 LOC100364002
Gene Name:
reproductive homeobox 9
reproductive homeobox 9-like
Gene Aliases:
-
-
Chromosome Location:
Chr.Un|NW_007906942: 9496 - 10900 on Build Rnor_6.0
Chromosome Location:
Chr.X: 123999804 - 124002065 on Build Rnor_6.0
UniGene ID:
Rn.144519
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 260851

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rhox9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024874.1 NP_001020045.1 4 645
GenBank mRNA DQ058656.1 - 4 645
Gene Symbol: LOC100364002
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257483.2 XP_006257545.1 4 637

Back To Top

More Information


Gene Ontology Categories:

Process(es)

DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header