This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC73 SIRNA Assays ›
Gene Symbol:
Cdc73
Gene Name:
cell division cycle 73
Gene Aliases:
Hrpt2
Chromosome Location:
Chr.13: 60403771 - 60496511 on Build Rnor_6.0
UniGene ID:
Rn.162332
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 260772

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc73
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024769.1 NP_001019940.1 16 1667
GenBank mRNA BC097445.1 - 16 1667 7456471

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145991, BC027756, AK080861, BC031127, AK048027.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
transcription, DNA-templated
mRNA polyadenylation
biological_process
negative regulation of cell proliferation
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of Wnt signaling pathway
positive regulation of mRNA 3'-end processing
protein destabilization
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
cellular response to lipopolysaccharide
negative regulation of G1/S transition of mitotic cell cycle
RNA polymerase II core binding
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0