This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NEURL3 SIRNA Assays ›
Gene Symbol:
Neurl3
Gene Name:
neuralized E3 ubiquitin protein ligase 3
Gene Aliases:
Lincr
Chromosome Location:
Chr.9: 42831737 - 42840628 on Build Rnor_6.0
UniGene ID:
Rn.17894
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 260438

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Neurl3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014100.1 NP_001014122.1 2 354
XM_008767025.1 XP_008765247.1 3 650
XM_008767026.1 XP_008765248.1 3 428
GenBank mRNA BC088198.1 - 2 354 7376146
FQ227363.1 - 2 419

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0