This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF346 SIRNA Assays ›
Gene Symbol:
ZNF346
Gene Name:
zinc finger protein 346
Gene Aliases:
JAZ, Zfp346
Chromosome Location:
Chr.5: 177022680 - 177081189 on Build GRCh38
UniGene ID:
Hs.484259
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 2604

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF346
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308214.1 NP_001295143.1 1, 2 238
NM_001308215.1 NP_001295144.1 1, 2 238
NM_001308216.1 NP_001295145.1 1, 2 238
NM_001308219.1 NP_001295148.1 1, 2 238
NM_012279.3 NP_036411.1 1, 2 238
NR_131773.1 1, 2 238
NR_131774.1 1, 2 238
XM_017009294.1 XP_016864783.1 1, 2 238
XM_017009295.1 XP_016864784.1 1, 2 238
XM_017009296.1 XP_016864785.1 1, 2 238
XM_017009297.1 XP_016864786.1 1, 2 238
XM_017009299.1 XP_016864788.1 1, 2 238
XM_017009301.1 XP_016864790.1 1, 2 238
GenBank mRNA AF083340.1 - 1, 2 211
AK026627.1 - 1, 2 206
AK295549.1 - 1, 2 214
AK300065.1 - 1, 2 202
AK300738.1 - 1, 2 183
AL359576.1 - 1, 2 186
BC007775.2 - 1, 2 193 4301820
BU860103.1 - 1, 2 213
BX331331.2 - 1, 2 193
CB988061.1 - 248
CR749692.1 - 1, 2 186
DA169130.1 - 1, 2 211
DA789054.1 - 1, 2 240

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107338, XM_006253628, FQ212045.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of apoptotic process
intrinsic apoptotic signaling pathway by p53 class mediator
p53 binding
double-stranded RNA binding
protein binding
zinc ion binding
enzyme binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0