This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIANP SIRNA Assays ›
Gene Symbol:
Pianp
Gene Name:
PILR alpha associated neural protein
Gene Aliases:
PANP, RGD1304952, leda-1
Chromosome Location:
Chr.4: 157509827 - 157517676 on Build Rnor_6.0
UniGene ID:
Rn.13411
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 260327

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pianp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014059.1 NP_001014081.1 5 1942
XM_006237370.2 XP_006237432.1 6 1626
XM_008763285.1 XP_008761507.1 6 3378
XM_008763286.1 XP_008761508.1 6 3418
GenBank mRNA BC085924.1 - 5 1942 7109219

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0