This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IL21R SIRNA Assays ›
Gene Symbol:
Il21r
Gene Name:
interleukin 21 receptor
Gene Aliases:
-
Chromosome Location:
Chr.1: 196996411 - 197024185 on Build Rnor_6.0
UniGene ID:
Rn.103248
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 259727

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il21r
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012469.1 NP_001012487.1 6 900
XM_006230261.2 XP_006230323.1 5 937
XM_006230262.2 XP_006230324.1 6 913
GenBank mRNA BC089857.1 - 6 900 7381492
FQ226709.1 - 5 771

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
cytokine-mediated signaling pathway
molecular_function
cytokine receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0