This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SENP8 SIRNA Assays ›
Gene Symbol:
Senp8
Gene Name:
SUMO/sentrin specific peptidase family member 8
Gene Aliases:
-
Chromosome Location:
Chr.8: 64558506 - 64572964 on Build Rnor_6.0
UniGene ID:
Rn.145873
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 259713

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Senp8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012355.1 NP_001012355.1 3 288
XM_006243196.2 XP_006243258.1 2 382
GenBank mRNA BC089939.1 - 3 288 7310036
FQ226292.1 - 2 270
FQ227853.1 - 3 301
FQ231841.1 - 2 280

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001172069, NM_001172071, NM_001172068, NM_027838, NM_001172070, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein deneddylation
proteolysis
NEDD8-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0