This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MGC108823 SIRNA Assays ›
Gene Symbol:
MGC108823 LOC102555392 LOC103694226
Gene Name:
similar to interferon-inducible GTPase
interferon-inducible GTPase 1-like
interferon-inducible GTPase 1-like
Gene Aliases:
Ifgga3
-
-
Chromosome Location:
Chr.18: 55576239 - 55584959 on Build Rnor_6.0
Chromosome Location:
Chr.18: 55530864 - 55532179 on Build Rnor_6.0
Chromosome Location:
Chr.18: 55466373 - 55467688 on Build Rnor_6.0
UniGene ID:
Rn.201788
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 259707

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MGC108823
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012353.1 NP_001012353.1 2 1326
GenBank mRNA BC089836.1 - 2 1326 7326302
Gene Symbol: LOC102555392
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006254757.2 XP_006254819.1 1208
Gene Symbol: LOC103694226
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772196.1 XP_008770418.1 1 1208

Back To Top

More Information


Gene Ontology Categories:

Process(es)

GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0