siRNA Details
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC081953.1 | - | 6 | 1337 | 7122794 |

Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC081953.1 | - | 6 | 1337 | 7122794 |