This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARHGAP24 SIRNA Assays ›
Gene Symbol:
Arhgap24
Gene Name:
Rho GTPase activating protein 24
Gene Aliases:
DR-NR#2
Chromosome Location:
Chr.14: 8383211 - 8510435 on Build Rnor_6.0
UniGene ID:
Rn.24657
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 259360

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arhgap24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012032.1 NP_001012032.1 1 308
GenBank mRNA BC085797.1 - 1 308 7109913

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK039617.

Gene Ontology Categories:

Function(s)

angiogenesis
signal transduction
cell differentiation
negative regulation of Rac protein signal transduction
wound healing, spreading of epidermal cells
negative regulation of ruffle assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0