This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WDR31 SIRNA Assays ›
Gene Symbol:
Wdr31
Gene Name:
WD repeat domain 31
Gene Aliases:
-
Chromosome Location:
Chr.5: 78305526 - 78324307 on Build Rnor_6.0
UniGene ID:
Rn.38938
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 259298

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdr31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011976.2 NP_001011976.2 11 1151
XM_006238237.2 XP_006238299.1 11 1115
XM_006238239.2 XP_006238301.1 11 1177
GenBank mRNA BC079286.1 - 12 1279 7134286

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_145241, NM_001012361, AK093240, AK313843, AL833635, and others.
This siRNA also targets the following transcripts in Mouse: NM_023597, AF274321, AK017259, AK141026, BC037102.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header