This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DCLK2 SIRNA Assays ›
Gene Symbol:
Dclk2
Gene Name:
doublecortin-like kinase 2
Gene Aliases:
CL2, CLICK-II, CLICK2, Dck2, RGD1308384
Chromosome Location:
Chr.2: 186116624 - 186248425 on Build Rnor_6.0
UniGene ID:
Rn.23327
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 259140

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dclk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009691.3 NP_001009691.3 18 3404
NM_001195832.1 NP_001182761.1 19 3417
GenBank mRNA AY673998.1 - 18 2876

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
intracellular signal transduction
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0