This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SF1 SIRNA Assays ›
Gene Symbol:
Sf1
Gene Name:
splicing factor 1
Gene Aliases:
Zfp162
Chromosome Location:
Chr.1: 221732616 - 221749220 on Build Rnor_6.0
UniGene ID:
Rn.1074
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 258890

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110793.1 NP_001104263.1 7 1100
NM_058210.2 NP_478117.2 7 1100
XM_006230641.2 XP_006230703.1 7 4001
XM_006230642.2 XP_006230704.1 7 4001
XM_006230643.2 XP_006230705.1 7 4001
XM_006230644.2 XP_006230706.1 7 4000
XM_006230645.2 XP_006230707.1 7 4000
XM_006230646.2 XP_006230708.1 6 3925
XM_006230647.2 XP_006230709.1 7 3999
XM_006230648.2 XP_006230710.1 7 3999
XM_008760065.1 XP_008758287.1 5 1914
XM_008760066.1 XP_008758288.1 7 4001
GenBank mRNA AF079873.1 - 4 380
BC081859.1 - 6 1013 7132476
FQ224273.1 - 7 1103

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

male sex determination
Leydig cell differentiation
negative regulation of smooth muscle cell proliferation
regulation of steroid biosynthetic process
RNA binding
zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0